2-Ethylpyridin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A17352.22
A17352.36
A17352.22EA
147
EUR
A17352.22
A17352.36
2-Ethylpyridin ≥98%
2-Ethylpyridine
Purity: 98%
CAS No.: 100-71-0
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 37°(98°F)
Refractive Index: 1.4970
Odour: Unpleasant
pH: 9-10
Melting Point: -63°
Boiling Point: 149°
Beilstein Reference: 106480
UN No.: 1993
MDL No.: MFCD00006361
Formel:
C₇H₉N Molecular Weight: 107,16 g/mol Siedepunkt: 149 °C Schmelzpunkt: –63 °C Dichte: 0,937 Flammpunkt: 37 °C (98 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006361 CAS-Nummer: 100-71-0 EINECS: 202-881-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...