2-Picolin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Methylpyridin, α-Picolin

A10847.0F A10847.AE A10847.AP
A10847.0FEA 123 EUR
A10847.0F A10847.AE A10847.AP
2-Picolin ≥98%
2-Picolin

Purity: 98+% CAS No.: 109-06-8 Molecular Formula: C6H7N Formula Weight: 93.13 Flash Point: 26°(78°F) Refractive Index: 1.5000 Melting Point: -70° Boiling Point: 128-129° Beilstein Reference: 104581 Merck Reference: 14,7400 UN No.: 2313 MDL No.: MFCD00006332

Formel: C₆H₇N
Molecular Weight: 93.13 g/mol
Siedepunkt: 128…129 °C
Schmelzpunkt: –70 °C
Dichte: 0.944
Flammpunkt: 26 °C (78 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006332
CAS-Nummer: 109-06-8
EINECS: 203-643-7
UN: 2313
ADR: 3 III
Merck Index: 12,07554

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR