3-Ethylpyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20318.06
B20318.14
B20318.06EA
55.1
EUR
B20318.06
B20318.14
3-Ethylpyridin ≥98%
3-Ethylpyridine
Purity: 98%
CAS No.: 536-78-7
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 49°(120°F)
Refractive Index: 1.5020
Odour: Unpleasant
Melting Point: -77°
Boiling Point: 165-166°
Beilstein Reference: 106479
Merck Reference: 14,3848
UN No.: 1993
MDL No.: MFCD00006413
Formel:
C₇H₉N Molecular Weight: 107.16 g/mol Siedepunkt: 165…166 °C Schmelzpunkt: –77 °C Dichte: 0.941 Flammpunkt: 49 °C (120 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006413 CAS-Nummer: 536-78-7 EINECS: 208-647-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...