2,3-Lutidin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,3-Dimethylpyridin
L03697.14
L03697.22
L03697.36
L03697.14EA
27.5
EUR
L03697.14
L03697.22
L03697.36
2,3-Lutidin ≥99%
2,3-Lutidin
Purity: 99%
CAS No.: 583-61-9
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 50°(122°F)
Refractive Index: 1.5080
Odour: Unpleasant
Melting Point: -17°
Boiling Point: 160-161°
Beilstein Reference: 106418
UN No.: 2929
MDL No.: MFCD00009605
Formel:
C₇H₉N Molecular Weight: 107,16 g/mol Siedepunkt: 160…161 °C Schmelzpunkt: –17 °C Dichte: 0,948 Flammpunkt: 50 °C (122 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009605 CAS-Nummer: 583-61-9 EINECS: 209-514-1 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...