3,5-Lutidin ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3,5-Dimethylpyridine

A14877.AC A14877.AE A14877.AP
A14877.ACEA 22.1 EUR
A14877.AC A14877.AE A14877.AP
3,5-Lutidin ≥99%
3,5-Lutidin

Purity: 99% CAS No.: 591-22-0 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 53°(127°F) Refractive Index: 1.5050 Odour: Unpleasant Melting Point: -9° Boiling Point: 169-170° Solubility: Sol Beilstein Reference: 105682 UN No.: 1992 MDL No.: MFCD00006404

Formel: C₇H₉N
Molecular Weight: 107,16 g/mol
Siedepunkt: 169…170 °C
Schmelzpunkt: –9 °C
Dichte: 0,941
Flammpunkt: 53 °C (127 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006404
CAS-Nummer: 591-22-0
EINECS: 209-708-6
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR