3-Chlorthiophen
Lieferant: Thermo Scientific
SB02032.10
SB02032.50
MAYBSB02032.10EA
0
EUR
MAYBSB02032.10
MAYBSB02032.50
3-Chlorthiophen
3-Chlorthiophen
Formel:
C₄H₃ClS Molecular Weight: 118.59 g/mol Siedepunkt: 133…135 °C (1013 hPa) Dichte: 1.276 g/cm³ (20 °C) |
MDL:
MFCD00043887 CAS-Nummer: 17249-80-8 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...