2,5-Diiodthiophen ≥99%

Lieferant: Thermo Scientific
Achtung

A12443.06 A12443.14
A12443.06EA 79.6 EUR
A12443.06 A12443.14
2,5-Diiodthiophen ≥99%
2,5-Diiodthiophen

Purity: 99% CAS No.: 625-88-7 Molecular Formula: C4H2I2S Formula Weight: 335.93 Storage Temperature: 2°C to 8°C Melting Point: 39-43° Boiling Point: 139-140°/15mm Beilstein Reference: 109915 MDL No.: MFCD00014525

Formel: C₄H₂I₂S
Molecular Weight: 335,93 g/mol
Siedepunkt: 139…140 °C (15 mmHg)
Schmelzpunkt: 39…43 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00014525
CAS-Nummer: 625-88-7
EINECS: 210-915-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR