Thiophen ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Thiofuran
A13941.0E
A13941.18
A13941.30
A13941.36
A13941.0EEA
294
EUR
A13941.0E
A13941.18
A13941.30
A13941.36
Thiophen ≥99%
Thiophen
Purity: 99%
CAS No.: 110-02-1
Molecular Formula: C4H4S
Formula Weight: 84.14
Flash Point: -9°(16°F)
Refractive Index: 1.5280
Odour: Stench
Melting Point: -38°
Boiling Point: 83-85°
Beilstein Reference: 103222
Merck Reference: 14,9353
UN No.: 2414
MDL No.: MFCD00005413
Formel:
C₄H₄S Molecular Weight: 84.14 g/mol Siedepunkt: 83…85 °C Schmelzpunkt: –38 °C Dichte: 1.066 Flammpunkt: –9 °C (16 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005413 CAS-Nummer: 110-02-1 EINECS: 203-729-4 UN: 2414 ADR: 3 II Merck Index: 12,09490 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...