2,3,5-Trimethylthiophen

Lieferant: CHEMPUR

FL18875.POR 018875-1G
CHMPFL18875.POREA 0 EUR
CHMPFL18875.POR FLUO018875-1G
2,3,5-Trimethylthiophen
2,3,5-Trimethylthiophene
Formel: C₇H₁₀S
MDL: MFCD00142784
CAS-Nummer: 1795-05-7
EINECS: 217-269-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR