1-Chloradamantan

Lieferant: CHEMPUR

JFD00559.250 JFD00559.1
CHMPJFD00559.250EA 0 EUR
CHMPJFD00559.250 MAYBJFD00559.1
1-Chloradamantan
1-Chloradamantan
Formel: C₁₀H₁₅Cl
Molecular Weight: 170,68 g/mol
Schmelzpunkt: 163…166 °C
Dichte: 1,26 g/cm³ (20 °C)
MDL: MFCD00075627
CAS-Nummer: 935-56-8
EINECS: 213-305-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR