1-Adamantanol 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Hydroxytricyclo[3.3.1.1(3,7)]decan, 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxyadamantan, Tricyclo[3.3.1.1(3,7)]decan-1-ol
157100050
157100250
157101000
ACRO157100050EA
19.8
EUR
ACRO157100050
ACRO157100250
ACRO157101000
1-Adamantanol 99%
1-Adamantanol
Formel:
C₁₀H₁₆O Molecular Weight: 152.24 g/mol Siedepunkt: 287 Schmelzpunkt: >240 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074729 CAS-Nummer: 768-95-6 EINECS: 212-202-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder or needles |
Infrared spectrum | Conforms |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...