1-Adamantanol ≥99%

Lieferant: Thermo Scientific

Synonyme: 1-Hydroxytricyclo[3.3.1.1(3,7)]decan, 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxyadamantan, Tricyclo[3.3.1.1(3,7)]decan-1-ol

A10209.09 A10209.18 A10209.30
A10209.09EA 28.7 EUR
A10209.09 A10209.18 A10209.30
1-Adamantanol ≥99%
1-Adamantanol

Purity: 99% CAS No.: 768-95-6 Molecular Formula: C10H16O Formula Weight: 152.24 Melting Point: ca 245° subl. Beilstein Reference: 1903952

Formel: C₁₀H₁₆O
Molecular Weight: 152,24 g/mol
Siedepunkt: 287 °C (1013 hPa)
Schmelzpunkt: ∼ 245 °C (Subl.)
Dichte: 1,06 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00074729
CAS-Nummer: 768-95-6
EINECS: 212-202-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR