di-Ammoniumthiosulfat, reinst

Lieferant: Bernd Kraft

Synonyme: AmmoniumthiosulfatAmmoniumhyposulfit

KRAF20082.6600EA 726 EUR
KRAF20082.6600 KRAF20082.5600 KRAF20082.3600
di-Ammoniumthiosulfat, reinst
di-Ammoniumthiosulfat
Formel: H₈N₂O₃S₂
Molecular Weight: 148,21 g/mol
Dichte: 1,67 g/cm³ (20 °C)
MDL: MFCD00015965
CAS-Nummer: 7783-18-8
EINECS: 231-982-0
Merck Index: 13,00565

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR