Kaliumchlorid 0.2 mol/l (0.2 N) aqueous solution
Lieferant: Bernd Kraft
Synonyme:
Potassiumchloride
12923.3000
12923.2000
12923.3000
12923.5000
XXXX12923.3000EA
0
EUR
XXXX12923.3000
KRAF12923.2000
KRAF12923.3000
KRAF12923.5000
Kaliumchlorid 0.2 mol/l (0.2 N) aqueous solution
Potassium chloride
Formel:
KCl Molecular Weight: 74.55 g/mol Siedepunkt: 1420 Schmelzpunkt: 773 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011360 CAS-Nummer: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...