tert-Amylethylether

Lieferant: ABCR

Synonyme: 1,1-Dimethylpropyl ethyl ether, Ethyl tert-pentyl ether

AB171118.0025 AB171118.0005
ABCRAB171118.0025EA 0 EUR
ABCRAB171118.0025 ABCRAB171118.0005
tert-Amylethylether
tert-Amylethylether
Formel: C₇H₁₆O
Molecular Weight: 116,2 g/mol
MDL: MFCD01727266
CAS-Nummer: 919-94-8
UN: 3271
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR