Methylpropionylacetat ≥99%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate
B24848.14
B24848.22
B24848.36
B24848.14EA
38.5
EUR
B24848.14
B24848.22
B24848.36
Methylpropionylacetat ≥99%
Methylpropionylacetat
Purity: 99%
CAS No.: 30414-53-0
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 71°(159°F)
Refractive Index: 1.4220
Boiling Point: 73-74°/5mm
Beilstein Reference: 1753684
MDL No.: MFCD00011705
Formel:
C₆H₁₀O₃ Molecular Weight: 130,14 g/mol Siedepunkt: 73…74 °C (5 mmHg) Dichte: 1,037 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011705 CAS-Nummer: 30414-53-0 EINECS: 250-184-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...