Methylpropionylacetat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate
023871-50G
023871-500G
FLUO023871-50GEA
50.3
EUR
FLUO023871-50G
FLUO023871-500G
Methylpropionylacetat
Methylpropionylacetat
Formel:
C₆H₁₀O₃ Molecular Weight: 130,14 g/mol |
MDL:
MFCD00011705 CAS-Nummer: 30414-53-0 EINECS: 250-184-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...