Cyclohexylphenylketon ≥98%
Lieferant: Thermo Scientific
B24816.06
B24816.14
B24816.22
B24816.06EA
28.5
EUR
B24816.06
B24816.14
B24816.22
Cyclohexylphenylketon ≥98%
Cyclohexylphenylketon
Purity: 98%
CAS No.: 712-50-5
Molecular Formula: C13H16O
Formula Weight: 188.27
Flash Point:
Formel:
C₁₃H₁₆O Molecular Weight: 188,27 g/mol Siedepunkt: 165…168 °C (20 mmHg) Schmelzpunkt: 52…57 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001467 CAS-Nummer: 712-50-5 EINECS: 211-923-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...