Ethyl-3,3-diethoxypropionat 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 3,3-diethoxypropanoate, Ethyl 3,3-diethoxypropionate

L09562.09 L09562.18
L09562.09EA 43.5 EUR
L09562.09 L09562.18
Ethyl-3,3-diethoxypropionat 96%
Ethyl-3,3-diethoxypropionat

Purity: 96% CAS No.: 10601-80-6 Molecular Formula: C9H18O4 Formula Weight: 190.24 Storage Temperature: -30°C to -10°C Flash Point: 93°(199°F) Refractive Index: 1.4120 Boiling Point: 92-93°/13mm Beilstein Reference: 1772110 MDL No.: MFCD00009865

Formel: C₉H₁₈O₄
Molecular Weight: 190.24 g/mol
Siedepunkt: 92…93 °C (13 mmHg)
Dichte: 0.978
Flammpunkt: 93 °C (199 °F)
MDL: MFCD00009865
CAS-Nummer: 10601-80-6
EINECS: 234-223-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR