5-Amino-2-methylbenzothiazol 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Methylbenzo[d]thiazol-5-amine, 2-Methyl-1,3-benzothiazol-5-amine

B23631.04 B23631.09
B23631.04EA 56.2 EUR
B23631.04 B23631.09
5-Amino-2-methylbenzothiazol 99%
5-Amino-2-methylbenzothiazole

Purity: 99% CAS No.: 13382-43-9 Molecular Formula: C8H8N2S Formula Weight: 164.22 MDL No.: MFCD00226291

Formel: C₈H₈N₂S
Molecular Weight: 164.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00226291
CAS-Nummer: 13382-43-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR