Dimethoxy(methyl)octadecylsilan

Lieferant: CHEMPUR SILANE

SILS12235.10 SILS12235.25 SILS12235.100
SILNSILS12235.10EA 0 EUR
SILNSILS12235.10 SILNSILS12235.25 SILNSILS12235.100
Dimethoxy(methyl)octadecylsilan
Dimethoxy(methyl)octadecylsilane
Formel: C₂₁H₄₆O₂Si
Molecular Weight: 358,68 g/mol
CAS-Nummer: 70851-50-2
EINECS: 274-936-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR