Dimethoxy(diisobutyl)silan

Lieferant: CHEMPUR SILANE

Synonyme: Diisobutyldimethoxysilane

SILS06675.10 SILS06675.50
SILNSILS06675.10EA 0 EUR
SILNSILS06675.10 SILNSILS06675.50
Dimethoxy(diisobutyl)silan
Dimethoxy(diisobutyl)silan
Formel: C₁₀H₂₄O₂Si
Molecular Weight: 204,38 g/mol
MDL: MFCD00236510
CAS-Nummer: 17980-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR