Ethylpentafluorpropionylacetat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 3-oxo-4,4,5,5,5-pentafluoropentanoate
002401-1G
002401-5G
FLUO002401-1GEA
31.1
EUR
FLUO002401-1G
FLUO002401-5G
Ethylpentafluorpropionylacetat
Ethylpentafluorpropionylacetat
Formel:
C₇H₇F₅O₃ Molecular Weight: 234,12 g/mol |
MDL:
MFCD00013569 CAS-Nummer: 663-35-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...