2,2-Difluorpropionsäure

Lieferant: FLUOROCHEM

Synonyme: 2,2-Difluoropropanoic acid

010408-1G 010408-5G
FLUO010408-1GEA 67.1 EUR
FLUO010408-1G FLUO010408-5G
2,2-Difluorpropionsäure
2,2-Difluorpropionsäure
Formel: C₃H₄F₂O₂
Molecular Weight: 110.06 g/mol
MDL: MFCD03093764
CAS-Nummer: 373-96-6
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR