1-Brom-4-phenylbutan ≥98%

Lieferant: Thermo Scientific

B25237.03 B25237.06
B25237.03EA 27.1 EUR
B25237.03 B25237.06
1-Brom-4-phenylbutan ≥98%
1-Brom-4-phenylbutan

Purity: 98% CAS No.: 13633-25-5 Molecular Formula: C10H13Br Formula Weight: 213.12 Boiling Point: 131-133°/12mm MDL No.: MFCD00154988

Formel: C₁₀H₁₃Br
Siedepunkt: 131…133 °C (12 mmHg)
Dichte: 1,214
Lagertemperatur: Raumtemperatur
MDL: MFCD00154988
CAS-Nummer: 13633-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR