Triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Triphenylphosphine hydrobromide

M-Clarity™ Qualitätsstufe = MQ200

338389-25G 338389-5G
SIAL338389-25GEA 88.4 EUR
SIAL338389-25G SIAL338389-5G
Triphenylphosphoniumbromid, Sigma-Aldrich®
Triphenylphosphonium bromide
Formel: C₁₈H₁₆BrP
Molecular Weight: 343,2 g/mol
MDL: MFCD00035107
CAS-Nummer: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR