Cyclohexylethylketon ≥99%

Lieferant: Thermo Scientific

B20409.06 B20409.14
B20409.06EA 192 EUR
B20409.06 B20409.14
Cyclohexylethylketon ≥99%
Cyclohexylethylketon

Purity: 99% CAS No.: 1123-86-0 Molecular Formula: C9H16O Formula Weight: 140.23 MDL No.: MFCD00060814

Formel: C₉H₁₆O
Dichte: 0,91
Lagertemperatur: Raumtemperatur
MDL: MFCD00060814
CAS-Nummer: 1123-86-0
EINECS: 214-380-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR