Cyclohexylmethylketon 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Cyclohexylethanone
B21000.14
B21000.06
B21000.22
B21000.14EA
117
EUR
B21000.14
B21000.06
B21000.22
Cyclohexylmethylketon 95%
Cyclohexylmethylketon
Purity: 95%
CAS No.: 823-76-7
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 66°(151°F)
Physical Form: Liquid
Density: 0.917
Refractive Index: 1.4520
Melting Point: -34°
Boiling Point: 181-183°
Beilstein Reference: 507229
MDL No.: MFCD00040418
Formel:
C₈H₁₄O Molecular Weight: 126.2 g/mol Siedepunkt: 181…183 °C Schmelzpunkt: –34 °C Dichte: 0.917 Flammpunkt: 66 °C (151 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040418 CAS-Nummer: 823-76-7 EINECS: 212-517-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...