(±)-Ethyl-2-brompropionat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate

M-Clarity™ Qualitätsstufe = MQ200

E14551-100G E14551-250G E14551-25G
SIALE14551-100GEA 48.2 EUR
SIALE14551-100G SIALE14551-250G SIALE14551-25G
(±)-Ethyl-2-brompropionat, Sigma-Aldrich®
(±)-Ethyl-2-brompropionat
Formel: C₅H₉BrO₂
Molecular Weight: 181,03 g/mol
MDL: MFCD00000144
CAS-Nummer: 535-11-5
EINECS: 208-609-5
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR