Oxetan ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Trimethylene oxide, 1,3-Trimethylene Oxide
A14452.06
A14452.14
A14452.22
A14452.06EA
97.4
EUR
A14452.06
A14452.14
A14452.22
Oxetan ≥97%
Oxetan
Purity: 97%
CAS No.: 503-30-0
Molecular Formula: C3H6O
Formula Weight: 58.08
Storage Temperature: 2°C to 8°C
Flash Point: -28°(-18°F)
Refractive Index: 1.3930
Melting Point: -97°
Boiling Point: 49-50°
Beilstein Reference: 102382
Merck Reference: 14,9715
UN No.: 1993
MDL No.: MFCD00005167
Formel:
C₃H₆O Molecular Weight: 58,08 g/mol Siedepunkt: 49…50 °C Schmelzpunkt: –97 °C Dichte: 0,891 Flammpunkt: –28 °C (–18 °F) |
MDL:
MFCD00005167 CAS-Nummer: 503-30-0 EINECS: 207-964-3 ADR: 3,II Merck Index: 14,9715 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...