Oxetan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethylene oxide, 1,3-Trimethylene Oxide
M-Clarity™ Qualitätsstufe = MQ200
T76201-10G
T76201-5G
SIALT76201-10GEA
229
EUR
SIALT76201-10G
SIALT76201-5G
Oxetan, Sigma-Aldrich®
Oxetan
Formel:
C₃H₆O Molecular Weight: 58,08 g/mol |
MDL:
MFCD00005167 CAS-Nummer: 503-30-0 EINECS: 207-964-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...