3-Azetidincarbonsäure ≥98%
Lieferant: Thermo Scientific
Synonyme:
3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid
H52294.03
H52294.06
H52294.03EA
120
EUR
H52294.03
H52294.06
3-Azetidincarbonsäure ≥98%
3-Azetidincarbonsäure
Purity: 98+%
CAS No.: 36476-78-5
Molecular Formula: C4H7NO2
Formula Weight: 101.11
Melting Point: ca 250° dec.
MDL No.: MFCD00191763
Formel:
C₄H₇NO₂ Molecular Weight: 101,11 g/mol Schmelzpunkt: ∼ 250 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191763 CAS-Nummer: 36476-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...