3-Azetidincarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid

MS-0192.1 MS-0192.5 MS-0192.500
BIOTMS-0192.1EA 0 EUR
BIOTMS-0192.1 BIOTMS-0192.5 BIOTMS-0192.500
3-Azetidincarbonsäure
3-Azetidincarbonsäure
Formel: C₄H₇NO₂
Molecular Weight: 101.11 g/mol
MDL: MFCD00191763
CAS-Nummer: 36476-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR