2-Imidazolidinthion, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione

M-Clarity™ Qualitätsstufe = MQ100

I504-100G I504-1KG
SIALI504-100GEA 58.1 EUR
SIALI504-100G SIALI504-1KG
2-Imidazolidinthion, Sigma-Aldrich®
2-Imidazolidinthion
Formel: C₃H₆N₂S
Molecular Weight: 102,16 g/mol
MDL: MFCD00005276
CAS-Nummer: 96-45-7
EINECS: 202-506-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR