Dimethoxy(methyl)silan

Lieferant: CHEMPUR SILANE

SILS11250.5 SILS11250.25 SILS11250.100
SILNSILS11250.5EA 0 EUR
SILNSILS11250.5 SILNSILS11250.25 SILNSILS11250.100
Dimethoxy(methyl)silan
Dimethoxy(methyl)silane
Formel: C₃H₁₀O₂Si
MDL: MFCD00048019
CAS-Nummer: 16881-77-9
EINECS: 240-914-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR