5-Brombenzotriazol

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-1H-1,2,3-benzotriazole, 5-Bromo-1H-benzotriazole

PY-1247-1G PY-1247-5G
COBBPY-1247-1GEA 235 EUR
COBBPY-1247-1G COBBPY-1247-5G
5-Brombenzotriazol
5-Bromobenzotriazole
Formel: C₆H₄BrN₃
Molecular Weight: 198,02 g/mol
MDL: MFCD00464510
CAS-Nummer: 32046-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR