(±)-Campherchinon ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
A14967.06 A14967.14 A14967.22
A14967.06EA 60.1 EUR
A14967.06 A14967.14 A14967.22
(±)-Campherchinon ≥99%
(±)-Campherchinon
Purity: 99% CAS No.: 10373-78-1 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 198-202° Beilstein Reference: 1909463
Formel: C₁₀H₁₄O₂
Molecular Weight: 166.22 g/mol
Schmelzpunkt: 198…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064160
CAS-Nummer: 10373-78-1
EINECS: 233-814-1