5-Brom-2-mercapto-6-methoxybenzothiazol

Lieferant: COMBI-BLOCKS

HC-3058-1G HC-3058-5G
COBBHC-3058-1GEA 204 EUR
COBBHC-3058-1G COBBHC-3058-5G
5-Brom-2-mercapto-6-methoxybenzothiazol
5-Bromo-2-mercapto-6-methoxybenzothiazole
Formel: C₈H₆BrNOS₂
MDL: MFCD15143549
CAS-Nummer: 1215205-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR