6-Brom-2-mercaptobenzothiazol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromobenzo[d]thiazole-2(3H)-thione

HI-1550-5G HI-1550-1G
COBBHI-1550-5GEA 154 EUR
COBBHI-1550-5G COBBHI-1550-1G
6-Brom-2-mercaptobenzothiazol
6-Bromo-2-mercaptobenzothiazole
Formel: C₇H₄BrNS₂
Molecular Weight: 246.15 g/mol
MDL: MFCD09880848
CAS-Nummer: 51618-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR