6-Fluor-2-mercaptobenzothiazol

Lieferant: COMBI-BLOCKS

Synonyme: 6-Fluorobenzo[d]thiazole-2(3H)-thione

HC-3053-5G HC-3053-1G
COBBHC-3053-5GEA 983 EUR
COBBHC-3053-5G COBBHC-3053-1G
6-Fluor-2-mercaptobenzothiazol
6-Fluoro-2-mercaptobenzothiazole
Formel: C₇H₄FNS₂
Molecular Weight: 185,25 g/mol
MDL: MFCD00665747
CAS-Nummer: 80087-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR