6-Ethoxy-2-mercaptobenzothiazol ≥99%

Lieferant: Thermo Scientific

Synonyme: 6-Ethoxybenzo[d]thiazole-2(3H)-thione

L04447.06 L04447.14
L04447.06EA 67 EUR
L04447.06 L04447.14
6-Ethoxy-2-mercaptobenzothiazol ≥99%
6-Ethoxy-2-mercaptobenzothiazole

Purity: 99% CAS No.: 120-53-6 Molecular Formula: C9H9NOS2 Formula Weight: 211.32 Melting Point: 196-201° Beilstein Reference: 152313

Formel: C₉H₉NOS₂
Molecular Weight: 211,31 g/mol
Schmelzpunkt: 196…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00467715
CAS-Nummer: 120-53-6
EINECS: 204-405-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR