4-Brom-2-mercapto-6-(trifluormethyl)benzothiazol

Lieferant: COMBI-BLOCKS

HC-3133-5G HC-3133-1G
COBBHC-3133-5GEA 595 EUR
COBBHC-3133-5G COBBHC-3133-1G
4-Brom-2-mercapto-6-(trifluormethyl)benzothiazol
4-Bromo-2-mercapto-6-(trifluoromethyl)benzothiazole
Formel: C₈H₃BrF₃NS₂
MDL: MFCD13195699
CAS-Nummer: 1215205-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR