(±)-2-Brompropionylchlorid 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Bromopropionylchloride

163620100 163620500 163622500
ACRO163620100EA 65.7 EUR
ACRO163620100 ACRO163620500 ACRO163622500
(±)-2-Brompropionylchlorid 98%
(±)-2-Brompropionylchlorid
Formel: C₃H₄BrClO
Molecular Weight: 171,42 g/mol
Siedepunkt: 131…133 °C (760 mmHg)
Dichte: 1,7 g/cm³
Flammpunkt: 51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000711
CAS-Nummer: 7148-74-5
EINECS: 230-466-2
UN: 2920
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4810 (20°C, 589 nm)
Titration Argentometric ≥97.5 % (ex Cl)
Phosphate (PO4) ≤0.5 ppm
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Bromine (Br) ≤50 ppm
Aluminium (Al) ≤0.05 ppm
Ammonium (NH4) ≤1 ppm
Arsenic (As) ≤0.01 ppm
Barium (Ba) ≤0.02 ppm
Beryllium (Be) ≤0.02 ppm
Bismuth (Bi) ≤0.1 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.02 ppm
Germanium (Ge) ≤0.05 ppm
Iron (Fe) ≤0.2 ppm
Lead (Pb) ≤0.02 ppm
Lithium (Li) ≤0.01 ppm
Magnesium (Mg) ≤0.01 ppm
Manganese (Mn) ≤0.1 ppm
Mercury (Hg) ≤0.01 ppm
Molybdenum (Mo) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Strontium (Sr) ≤0.01 ppm
Thallium (Tl) ≤0.05 ppm
Titanium (Ti) ≤0.1 ppm
Vanadium (V) ≤0.01 ppm
Zinc (Zn) ≤0.05 ppm
Zirconium (Zr) ≤0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR