(±)-2-Brompropionylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Bromopropionylchloride

B23662.09 B23662.18
B23662.09EA 65 EUR
B23662.09 B23662.18
(±)-2-Brompropionylchlorid ≥98%
(±)-2-Brompropionylchlorid

Purity: 98% CAS No.: 7148-74-5 Molecular Formula: CH3CHBrCOCl Formula Weight: 171.43 Flash Point: 51°(125°F) Refractive Index: 1.4780 Boiling Point: 131-133° Beilstein Reference: 773800 UN No.: 2920 MDL No.: MFCD00000711

Formel: C₃H₄BrClO
Molecular Weight: 171,42 g/mol
Siedepunkt: 131…133 °C
Dichte: 1,700
Flammpunkt: 51 °C (125 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000711
CAS-Nummer: 7148-74-5
EINECS: 230-466-2
UN: 2920
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR