(±)-2-Chlorpropionylchlorid, Sigma-Aldrich®

Lieferant: Merck


MQ100

157139-100G 157139-25G
SIAL157139-100GEA 120 EUR
SIAL157139-100G SIAL157139-25G
(±)-2-Chlorpropionylchlorid, Sigma-Aldrich®
(±)-2-Chlorpropionylchlorid
Formel: C₃H₄Cl₂O
Molecular Weight: 126.97 g/mol
MDL: MFCD00000862
CAS-Nummer: 7623-09-8
EINECS: 231-540-7
UN: 2920
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR