(±)-Methyl-2,3-dichlorpropionat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl 2,3-dichloropropanoate
B21993.14
B21993.22
B21993.36
B21993.14EA
35.4
EUR
B21993.14
B21993.22
B21993.36
(±)-Methyl-2,3-dichlorpropionat ≥98%
(±)-Methyl-2,3-dichlorpropionat
Purity: 98%
CAS No.: 3674-09-7
Molecular Formula: C4H6Cl2O2
Formula Weight: 157.00
Flash Point: 87°(188°F)
Refractive Index: 1.4530
Boiling Point: 91-92°/50mm
Beilstein Reference: 1701891
UN No.: 3265
MDL No.: MFCD00000944
Formel:
C₄H₆Cl₂O₂ Molecular Weight: 157 g/mol Siedepunkt: 91…92 °C (50 mmHg) Dichte: 1,326 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000944 CAS-Nummer: 3674-09-7 EINECS: 222-940-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...