Methylcarbazat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl hydrazinecarboxylate, Methyl hydrazinocarboxylate

166141000 166145000 166140025
ACRO166141000EA 49.2 EUR
ACRO166141000 ACRO166145000 ACRO166140025
Methylcarbazat 97%
Methylcarbazat
Formel: C₂H₆N₂O₂
Molecular Weight: 90,08 g/mol
Siedepunkt: 108 °C (12 mmHg)
Schmelzpunkt: 68…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007594
CAS-Nummer: 6294-89-9
EINECS: 228-560-3
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to pink Crystalline powder or crystals or platelets
Infrared spectrum Conforms
Melting point 68 to 73 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR