Formamidinacetat 99%

Lieferant: Thermo Scientific
Achtung

119711000 119715000
ACRO119711000EA 49.3 EUR
ACRO119711000 ACRO119715000
Formamidinacetat 99%
Formamidinacetat
Formel: C₃H₈N₂O₂
Molecular Weight: 104,11 g/mol
Schmelzpunkt: 157…161 °C
Dichte: 1,124 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00012866
CAS-Nummer: 3473-63-0
EINECS: 222-442-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR