Formamidinacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

F15803-100G F15803-500G
SIALF15803-100GEA 57.2 EUR
SIALF15803-100G SIALF15803-500G
Formamidinacetat, Sigma-Aldrich®
Formamidinacetat
Formel: C₃H₈N₂O₂
Molecular Weight: 104.11 g/mol
MDL: MFCD00012866
CAS-Nummer: 3473-63-0
EINECS: 222-442-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR