S-Methylthioacetat ≥98%

Lieferant: Thermo Scientific
Gefahr

A12400.06 A12400.14 A12400.22
A12400.06EA 48.7 EUR
A12400.06 A12400.14 A12400.22
S-Methylthioacetat ≥98%
S-Methylthioacetat

Purity: 98+% CAS No.: 1534-08-3 Molecular Formula: C3H6OS Formula Weight: 91.14 Flash Point: 12°(54°F) Odour: Unpleasant Boiling Point: 97-99° UN No.: 1993 MDL No.: MFCD00014989

Formel: C₃H₆OS
Molecular Weight: 90,15 g/mol
Siedepunkt: 97…99 °C
Dichte: 1,024
Flammpunkt: 12 °C (54 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014989
CAS-Nummer: 1534-08-3
EINECS: 216-252-1
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR